To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD40 Molecule
- Antigen Symbol:CD40
- Clonality:Monoclonal
- Clone:4F11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:958
- Antigen Synonyms:TNF receptor superfamily member 5|TRAP|AI326936|p50|IMD3|GP39|Tnfrsf5|IGM|T-BAM|Bp50|HIGM1
- Amino Acid Number:21 to 193
- Storage Buffer:1x PBS, pH 7.4
- Sequence:EPPTACREKQYLINSQCCSLCQPGQKLVSDCTEFTETECLPCGESEFLDTWNRETHCHQHKYCDPNLGLRVQQKGTSETDTICTCEEGWHCTSEACESCVLHRSCSPGFGVKQIATGVSDTICEPCPVGFFSNVSSAFEKCHPWTSCETKDLVVQQAGTNKTDVVCGPQDRLR
- Storage Temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:CD40 (AAH12419, 21 a.a. ~ 193 a.a) partial recombinant protein with GST tag.
- Cat. No.:10594-328
- Supplier no.:H00000958-M26
Specifications
About this item
Mouse monoclonal antibody raised against a partial recombinant CD40.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: