Anti-Newcastle disease virus Mouse Monoclonal Antibody [clone: 9F7]
Catalog # 10676-894
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Newcastle Disease Virus
- Antigen Symbol:NDV
- Clonality:Monoclonal
- Clone:9F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Size:200 µg
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:PBS, pH 7.4 (0.1% sodium azide)
- Storage Temperature:Store at 4°C.
- Immunogen:Newcastle disease virus (NDV), La-sota strain.
- Cat. No.:10676-894
- Supplier no.:MAB0572
Specifications
About this item
Mouse monoclonal antibody raised against Newcastle disease virus, La-sota strain.
Type: Primary
Antigen: Newcastle disease virus
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 9F7
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: