Anti-CDK9 Mouse Monoclonal Antibody [clone: 1B5A7]
Catalog # 10707-336
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Cyclin-dependent Kinase 9
- Antigen Symbol:CDK9
- Clonality:Monoclonal
- Clone:1B5A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:1025
- Antigen Synonyms:CDC2-related kinase|C-2k|tat-associated kinase complex catalytic subunit|CTK1|serine/threonine-protein kinase PITALRE|Cell Division Cycle 2-like protein kinase 4|cell division protein kinase 9|PITALRE|TAK|serine/threonine protein kinase PITALRE|CDC2L4|RP11-228B15.5
- Storage Buffer:ascitic (0.03% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human CDK9.
- Cat. No.:10707-336
- Supplier no.:MAB10467
Specifications
About this item
Mouse monoclonal antibody raised against recombinant CDK9.
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; ELISA: 1:10000
Type: Primary
Antigen: CDK9
Clonality: Monoclonal
Clone: 1B5A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: