Anti-NCR1 Mouse Monoclonal Antibody [clone: n1D9]
10726-776
: Abnova
- Antibody Type:Primary
- Antigen Name:Natural Cytotoxicity Triggering Receptor 1
- Antigen Symbol:NCR1
- Clonality:Monoclonal
- Clone:n1D9
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:9437
- Antigen Synonyms:CD335|NK-p46|LY94|NKP46
- Amino Acid Number:22 to 255
- Storage Buffer:PBS, pH 7.4 (0.1% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 22-255 of human NCR1.
- Cat. No.:10726-776
- Supplier no.:MAB1052
Mouse monoclonal antibody raised against partial recombinant NCR1.
Type: Primary
Antigen: NCR1
Clonality: Monoclonal
Clone: n1D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human