Anti-DMD Mouse Monoclonal Antibody [clone: DYS-48]
Catalog # 10710-926
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Dystrophin
- Antigen Symbol:DMD
- Clonality:Monoclonal
- Clone:DYS-48
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Chicken,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:1756
- Antigen Synonyms:DXS230|MRX85|DXS164|BMD|DXS272|DXS268|DXS206|DXS142|CMD3B|DXS269|DXS270|DXS239
- Storage Buffer:1.2% sodium acetate (2 mg BSA, 0.01 mg sodium azide)
- Storage Temperature:Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human DMD.
- Purification:Affinity purification
- Cat. No.:10710-926
- Supplier no.:MAB1453
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant DMD.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml
Type: Primary
Antigen: DMD
Clonality: Monoclonal
Clone: DYS-48
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Chicken, Human, Mouse, Rat