To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cathepsin D
- Antigen Symbol:CTSD
- Clonality:Monoclonal
- Clone:CP-D13
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:1509
- Antigen Synonyms:lysosomal aspartyl protease|HEL-S-130P|ceroid-lipofuscinosis|lysosomal aspartyl peptidase|CPSD|CLN10|neuronal 10
- Storage Buffer:1.2% sodium acetate (2 mg BSA, 0.01 mg sodium azide)
- Storage Temperature:Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human CTSD.
- Purification:Affinity purification
- Cat. No.:10709-406
- Supplier no.:MAB1513
Specifications
About this item
Mouse monoclonal antibody raised against native CTSD.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-2 ?g/ml
Type: Primary
Antigen: CTSD
Clonality: Monoclonal
Clone: CP-D13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: