Anti-EPHA2 Mouse Monoclonal Antibody [clone: 3D7]
10712-208
: Abnova
- Antibody Type:Primary
- Antigen Name:Ephrin type A receptor 2
- Antigen Symbol:EPHA2
- Clonality:Monoclonal
- Clone:3D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:1969
- Antigen Synonyms:Epithelial cell kinase|CTPA|ARCC2|ECK|CTRCT6|CTPP1|Ephrin type-A receptor 2|Tyrosine-protein kinase receptor ECK
- Amino Acid Number:559 to 976
- Storage Buffer:PBS, pH 7.4 (0.1% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 559-976 of human EPHA2.
- Cat. No.:10712-208
- Supplier no.:MAB1769
Mouse monoclonal antibody raised against partial recombinant EPHA2.
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunohistochemistry: 1:100-1:200
Type: Primary
Antigen: EPHA2
Clonality: Monoclonal
Clone: 3D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human