Order Entry
United States
ContactUsLinkComponent
Anti-XPO4 Rabbit Polyclonal Antibody
Anti-XPO4 Rabbit Polyclonal Antibody
Catalog # 10744-072
Supplier:  Abnova
undefined
Anti-XPO4 Rabbit Polyclonal Antibody
Catalog # 10744-072
Supplier:  Abnova
Supplier Number:  PAB0297

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Exportin 4
  • Antigen Symbol:
    XPO4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Monkey,
    Chicken,
    Chimpanzee,
    Human,
    Dog,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    64328
  • Amino Acid Number:
    850 to 950
  • Storage Buffer:
    PBS (0.2% gelatin, 0.05% sodium azide)
  • Storage Temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids 850-950 of human XPO4.
  • Cat. No.:
    10744-072
  • Supplier no.:
    PAB0297

Specifications

About this item

Rabbit polyclonal antibody raised against partial recombinant XPO4.

Type: Primary
Antigen: XPO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Chimpanzee, Dog, Human, Monkey, Mouse, Rat