Anti-EDEM3 Rabbit Polyclonal Antibody
10713-390
: Abnova
- Antibody Type:Primary
- Antigen Name:ER degradation enhancer, mannosidase alpha-like 3
- Antigen Symbol:EDEM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:80267
- Antigen Synonyms:C1orf22
- Storage Buffer:PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
- Sequence:YLYLLFADKEDIIFDIEDYIFTTEAHLLPLWLSTTNQSISKKNTTSEYTELDDSNFDWTCPNTQILFPNDPLYAQSIREPL
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human EDEM3.
- Purification:Antigen affinity purification
- Cat. No.:10713-390
- Supplier no.:PAB21781
Rabbit polyclonal antibody raised against recombinant EDEM3.
Recommended Dilutions: Immunohistochemistry: 1:10-1:20
Type: Primary
Antigen: EDEM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human