To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Adeno-associated Virus VP1/VP2/VP3
- Antigen Symbol:AAV VP1/VP2/VP3
- Clonality:Monoclonal
- Clone:B1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Virus
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Format:Lyophilized
- Storage Buffer:PBS (0.5%BSA)
- Storage Temperature:Store at 4 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Adeno-associated virus capsid proteins and virus particles.
- Cat. No.:10684-362
- Supplier no.:MAB4645
Specifications
About this item
Mouse monoclonal antibody raised against Adeno-associated Virus (AAV), VP1, VP2 and VP3.
Recommended Dilutions: Immunohistochemistry: 1:10
Type: Primary
Antigen: AAV VP1/VP2/VP3
Clonality: Monoclonal
Clone: B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus