Anti-HSF1 Rat Monoclonal Antibody [clone: 10H8]
Catalog # 10717-878
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Heat Shock Transcription Factor 1
- Antigen Symbol:HSF1
- Clonality:Monoclonal
- Clone:10H8
- Conjugation:Unconjugated
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Hamster,Monkey,Birds,Human,Bovine,Guinea pig,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:15499
- Antigen Synonyms:HSTF 1|Heat shock transcription factor 1|Heat shock factor protein 1|HSTF1|HSF 1
- Storage Buffer:PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to mouse Hsf1.
- Cat. No.:10717-878
- Supplier no.:MAB2285
Specifications
About this item
Rat monoclonal antibody raised against recombinant Hsf1.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunocytochemistry: 10 ?g/ml; Immunoprecipitation: 12.5 ?g/ml
Type: Primary
Antigen: HSF1
Clonality: Monoclonal
Clone: 10H8
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Birds, Bovine, Guinea Pig, Hamster, Human, Monkey, Mouse, Rat