Anti-SLITRK5 Rabbit Polyclonal Antibody
Catalog # 10738-040
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:SLIT And NTRK-like Family member 5
- Antigen Symbol:SLITRK5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:26050
- Antigen Synonyms:bA364G4.2|SLIT and NTRK-like protein 5|Slitrk5|KIAA0918|Leucine-rich repeat-containing protein 11|LRRC11
- Storage Buffer:PBS (0.02% sodium azide)
- Storage Temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal region 14 amino acids of human SLITRK5.
- Cat. No.:10738-040
- Supplier no.:PAB13334
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of SLITRK5.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: SLITRK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat