Anti-TMEM261 Rabbit Polyclonal Antibody
Catalog # 10704-522
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Transmembrane Protein 261
- Antigen Symbol:TMEM261
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:90871
- Antigen Synonyms:C9orf123
- Storage Buffer:PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
- Sequence:MGSRLSQPFESYITAPPGTAAAPAKPAPPATPGAPTSPAEHRLLKTCWSC
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human C9orf123.
- Purification:Antigen affinity purification
- Cat. No.:10704-522
- Supplier no.:PAB20745
Specifications
About this item
Rabbit polyclonal antibody raised against recombinant C9orf123.
Type: Primary
Antigen: TMEM261
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human