Anti-immunodeficiency virus type 1 gp41 Mouse Monoclonal Antibody [clone: 5A1]
10717-448
: Abnova
- Antibody Type:Primary
- Antigen Name:immunodeficiency virus type 1 gp41
- Antigen Symbol:immunodeficiency virus type 1 gp41
- Clonality:Monoclonal
- Clone:5A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Virus
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Amino Acid Number:466 to 753
- Storage Buffer:PBS, pH 7.2 (0.01% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 466-753 of HIV-1 gp41.
- Cat. No.:10717-448
- Supplier no.:MAB4809
Mouse monoclonal antibody raised against partial recombinant human immunodeficiency virus type 1 gp41.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: immunodeficiency virus type 1 gp41
Clonality: Monoclonal
Clone: 5A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Viruses