Anti-MMP3 Mouse Monoclonal Antibody [clone: SB14d]
Catalog # 10725-652
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Matrix Metalloproteinase 3
- Antigen Symbol:MMP3
- Clonality:Monoclonal
- Clone:SB14d
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:4314
- Antigen Synonyms:STR1|transin-1|MMP-3|proteoglycanase|CHDS6|STMY|SL-1|matrix metalloproteinase 3|STMY1
- Storage Buffer:100 mM BBS, pH 8.2
- Storage Temperature:Store at 4°C.
- Immunogen:Recombinant protein corresponding to human MMP3.
- Cat. No.:10725-652
- Supplier no.:MAB3331
Specifications
About this item
Mouse monoclonal antibody raised against recombinant MMP3.
Recommended Dilutions: ELISA: 1:3000-1:8000; Immunohistochemistry: 2 ?g/ml
Type: Primary
Antigen: MMP3
Clonality: Monoclonal
Clone: SB14d
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human