Anti-RBM8A Mouse Monoclonal Antibody [clone: 4C4]
Catalog # 10731-308
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:RNA Binding Motif Protein 8A
- Antigen Symbol:RBM8A
- Clonality:Monoclonal
- Clone:4C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human,Xenopus laevis
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:9939
- Antigen Synonyms:Y14
- Storage Buffer:PBS (0.1% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Human RBM8A.
- Cat. No.:10731-308
- Supplier no.:MAB2484
Specifications
About this item
Mouse monoclonal antibody raised against RBM8A.
Type: Primary
Antigen: RBM8A
Clonality: Monoclonal
Clone: 4C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Clawed frog, Human