To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Glycoprotein Ib (platelet), Alpha Polypeptide
- Antigen Symbol:GP1BA
- Clonality:Monoclonal
- Clone:SZ2
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:200 µg
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:2811
- Antigen Synonyms:GP1B|CD42b-alpha|CD42B|BSS|VWDP|BDPLT3|GPIbA|BDPLT1|DBPLT3
- Storage Buffer:PBS (1mg/mL BSA)
- Storage Temperature:Store at 4 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified GP1BA from human washed platelets.
- Cat. No.:10676-320
- Supplier no.:MAB4144
Specifications
About this item
Mouse monoclonal antibody raised against native GP1BA.
Recommended Dilutions: Flow Cytometry: 2?g/5x10^6 platelets/test)
Type: Primary
Antigen: GP1BA
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: SZ2
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human