Anti-NPHS1 Rabbit Polyclonal Antibody
Catalog # 10727-558
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Nephrin
- Antigen Symbol:NPHS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:4868
- Antigen Synonyms:Renal glomerulus-specific cell adhesion receptor|NPHN|CNF|SAPAP1
- Storage Buffer:PBS (0.02% sodium azide)
- Storage Temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus 14 amino acids of human NPHS1.
- Cat. No.:10727-558
- Supplier no.:PAB12848
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of NPHS1.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: NPHS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat