Anti-Col5a1 Rabbit Polyclonal Antibody
Catalog # 10735-300
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Symbol:Col5a1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Rat
- Size:500 µL
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:85490|85250|60379
- Storage Temperature:Store at 4 °C on dry atmosphere. After reconstitution with 0.5 mL distilled water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified Col5a1/Col5a2/Col5a3 from rat skin.
- Cat. No.:10735-300
- Supplier no.:PAB13509
Specifications
About this item
Rabbit polyclonal antibody raised against native Col5a1/Col5a2/Col5a3.
Recommended Dilutions: Immunohistochemistry: 1:200-1:600; Immunofluorescence: 1:40
Type: Primary
Antigen: Col5a1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: