To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Tumor Protein p53-binding Protein 1
- Antigen Symbol:TP53BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:7158
- Antigen Synonyms:p53-binding protein 1|tumor protein 53-binding protein|tumor suppressor p53-binding protein 1|53BP1|p53BP1|p202
- Storage Buffer:Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
- Storage Temperature:Store at 4°C. Do not freeze.
- Immunogen:Recombinant protein corresponding to human TP53BP1.
- Cat. No.:10742-490
- Supplier no.:PAB12506
Specifications
About this item
Rabbit polyclonal antibody raised against partial recombinant TP53BP1.
Recommended Dilutions: Immunocytochemistry: 1:100; Immunoprecipitation: 2-10 mg/mg lysate; Western Blot: 1:5000-1:25000
Type: Primary
Antigen: TP53BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse