Anti-FANCD2 Rabbit Polyclonal Antibody
10677-702
: Abnova
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Fanconi Anemia Complementation Group D2
- Antigen Symbol:FANCD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:200 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:2177
- Antigen Synonyms:FAD|FA4|FANCD|FACD|FA-D2|FAD2
- Modification:phospho-Ser222
- Storage Buffer:No additive
- Storage Temperature:Store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S222 of human FANCD2.
- Cat. No.:10677-702
- Supplier no.:PAB12530
Rabbit polyclonal antibody raised against synthetic phosphopeptide of FANCD2.
Type: Primary
Antigen: FANCD2
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human