Anti-Tubulin-alpha Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TU-01]
10688-870
: Abnova
- Antibody Type:Primary
- Antigen Name:Tubulin alpha
- Antigen Symbol:TUBA
- Clonality:Monoclonal
- Clone:TU-01
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Pig
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Antigen Synonyms:tubulin alpha-3 chain|tubulin B-alpha-1|alpha|tubulin|hum-a-tub1|alpha-tubulin 3|alpha 3|brain-specific|tubulin alpha-1A chain|hum-a-tub2
- Storage Buffer:PBS, pH 7.4 (15 mM sodium azide)
- Storage Temperature:Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified porcine tubulin-alpha.
- Cat. No.:10688-870
- Supplier no.:MAB5027
Mouse monoclonal antibody raised against native tubulin-alpha.
Recommended Dilutions: Immunofluorescence: 1:50
Type: Primary
Antigen: Tubulin-alpha
Clonality: FITC (Fluorescein Isothiocyanate)
Clone: Monoclonal
Conjugation: TU-01
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Pig