Order Entry
United States
ContactUsLinkComponent
Anti-HLA-G Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-G/9]
Anti-HLA-G Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-G/9]
Catalog # 10717-586
Supplier:  Abnova
undefined
Anti-HLA-G Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-G/9]
Catalog # 10717-586
Supplier:  Abnova
Supplier Number:  MAB5056

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    HLA class I histocompatibility antigen, alpha chain G
  • Antigen Symbol:
    HLA-G
  • Clonality:
    Monoclonal
  • Clone:
    MEM-G/9
  • Conjugation:
    APC (Allophycocyanin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3135
  • Antigen Synonyms:
    HLA class I histocompatibility antigen|DADB-15K14.8|alpha chain G|HLA-G histocompatibility antigen|b2 microglobulin|MHC-G|HLA class I molecule|HLA G antigen|MHC class I antigen G|class I|G
  • Storage Buffer:
    PBS (0.2% BSA, 15 mM sodium azide)
  • Storage Temperature:
    Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human HLA-G refolded with beta2-microglobulin and peptide.
  • Cat. No.:
    10717-586
  • Supplier no.:
    MAB5056

Specifications

About this item

Mouse monoclonal antibody raised against recombinant HLA-G.

Recommended Dilutions: Flow Cytometry: 1:100

Type: Primary
Antigen: HLA-G
Clonality: Monoclonal
Clone: MEM-G/9
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human