Order Entry
United States
ContactUsLinkComponent
Anti-MST1R Rabbit Polyclonal Antibody
Anti-MST1R Rabbit Polyclonal Antibody
Catalog # 10675-000
Supplier:  Abnova
undefined
Anti-MST1R Rabbit Polyclonal Antibody
Catalog # 10675-000
Supplier:  Abnova
Supplier Number:  PAB12613

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Macrophage Stimulating 1 Receptor
  • Antigen Symbol:
    MST1R
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    4486
  • Antigen Synonyms:
    soluble R|RON variant E2E3|MST1R variant RON30|PTK8|macrophage-stimulating protein receptor|MST1R variant RON62|soluble RON variant 1|c-met-related tyrosine kinase|CDw136|PTK8 protein tyrosine kinase 8|MSP receptor|p185-Ron|soluble RON variant 3|RON|soluble RON variant 2|CD136
  • Modification:
    phospho-Tyr1239
  • Storage Buffer:
    TBS, pH 7.2 (10% Proclin300)
  • Storage Temperature:
    Store at 4 °C. For long term storage store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Synthetic phosphopeptide corresponding to residues surrounding Y1239 of human MST1R.
  • Cat. No.:
    10675-000
  • Supplier no.:
    PAB12613

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic phosphopeptide of MST1R.

Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml; Immunohistochemistry: 2-5 ?g/ml

Type: Primary
Antigen: MST1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat