Anti-MAP3K12 Rabbit Polyclonal Antibody
Catalog # 10679-204
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Mitogen-activated Protein Kinase Kinase Kinase 12
- Antigen Symbol:MAP3K12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:7786
- Antigen Synonyms:MEKK12|ZPK|MUK|ZPKP1|DLK
- Storage Buffer:PBS (0.09% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human MAP3K12.
- Purification:Protein G purification
- Cat. No.:10679-204
- Supplier no.:PAB2777
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MAP3K12.
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500; Immunohistochemistry: 1:50-100
Type: Primary
Antigen: MAP3K12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse