Anti-ACRV1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Hs-14]
Catalog # 10683-138
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Acrosomal Vesicle Protein 1
- Antigen Symbol:ACRV1
- Clonality:Monoclonal
- Clone:Hs-14
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 Reactions
- Cross Adsorption:No
- Format:Liquid
- Gene ID:56
- Antigen Synonyms:D11S4365|SP-10|SPACA2
- Storage Buffer:PBS (0.2% BSA, 15 mM sodium azide)
- Storage Temperature:Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human ACRV1.
- Cat. No.:10683-138
- Supplier no.:MAB4490
Specifications
About this item
Mouse monoclonal antibody raised against native ACRV1.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: ACRV1
Clonality: Monoclonal
Clone: Hs-14
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse