Anti-DACT3 Mouse Monoclonal Antibody [clone: 2A5]
Catalog # 10710-160
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:dishevelled-binding antagonist of beta-catenin 3
- Antigen Symbol:DACT3
- Clonality:Monoclonal
- Clone:2A5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:147906
- Antigen Synonyms:RRR1|DAPPER3
- Amino Acid Number:89 to 202
- Storage Buffer:PBS, pH 7.4 (0.1% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 89-202 of human DACT3.
- Cat. No.:10710-160
- Supplier no.:MAB2064
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant DACT3.
Type: Primary
Antigen: DACT3
Clonality: Monoclonal
Clone: 2A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse