To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:T Cell Receptor Associated Transmembrane Adaptor 1
- Antigen Symbol:TRAT1
- Clonality:Monoclonal
- Clone:TRIM-04
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:50852
- Antigen Synonyms:TCRIM|TRIM|HSPC062
- Storage Buffer:PBS (0.2% BSA, 15 mM sodium azide)
- Storage Temperature:Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to partial human TRAT1.
- Cat. No.:10742-738
- Supplier no.:MAB4526
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant TRAT1.
Recommended Dilutions: Flow Cytometry: 1:50
Type: Primary
Antigen: TRAT1
Clonality: Monoclonal
Clone: TRIM-04
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human