Anti-CCL3 Mouse Monoclonal Antibody [clone: 199]
Catalog # 10734-688
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Chemokine (C-C motif) Ligand 3
- Antigen Symbol:CCL3
- Clonality:Monoclonal
- Clone:199
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:500 µg
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:6348
- Antigen Synonyms:G0S19-1|SCYA3|LD78ALPHA|MIP-1-alpha|MIP1A|C-C motif chemokine ligand 3
- Storage Buffer:10 mM PBS, pH 7.2
- Storage Temperature:Store at –20 °C. Aliquot after reconstitution to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human CCL3.
- Cat. No.:10734-688
- Supplier no.:MAB6299
Specifications
About this item
Mouse monoclonal antibody raised against recombinant CCL3.
Type: Primary
Antigen: CCL3
Clonality: Monoclonal
Clone: 199
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human