Anti-LIME Mouse Monoclonal Antibody [clone: mLIME-05]
Catalog # 10723-656
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Lck interacting transmembrane adaptor 1
- Antigen Symbol:LIME
- Clonality:Monoclonal
- Clone:mLIME-05
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:72699
- Antigen Synonyms:lck-interacting membrane protein|dJ583P15.4|LIME|LP8067|lck-interacting molecule|RP4-583P15.5|FLJ20406|lck-interacting transmembrane adapter 1
- Amino Acid Number:86 to 202
- Storage Buffer:PBS, pH 7.4 (15 mM sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 86-202 of mouse Lime1.
- Cat. No.:10723-656
- Supplier no.:MAB8509
Specifications
About this item
Mouse monoclonal antibody raised against partial recombiant Lime1.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: LIME
Clonality: Monoclonal
Clone: mLIME-05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse