Anti-ITGAV Mouse Monoclonal Antibody [clone: P2W7]
Catalog # 10721-532
Supplier: Abnova
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Integrin Alpha 5
- Antigen Symbol:ITGA5
- Clonality:Monoclonal
- Clone:P2W7
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:3685
- Antigen Synonyms:MSK8|Vitronectin Receptor Subunit Alpha|CD51|ITGAV|Vitronectin Receptor|VNRA|VTNR
- Storage Buffer:50 mM sodium phosphate buffer, 100 mM potassium Chloride, 150 mM NaCl, pH 7.5 (0.5 mg/mL gentamicsulfate)
- Storage Temperature:Store at 4°C.
- Immunogen:Native purified ITGAV from human ocular melanoma cell line (V+B2).
- Cat. No.:10721-532
- Supplier no.:MAB6943
Specifications
About this item
Mouse monoclonal antibody raised against native ITGAV.
Type: Primary
Antigen: ITGAV
Clonality: Monoclonal
Clone: P2W7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human