Anti-PPP2R3A Goat Polyclonal Antibody
Catalog # 10733-740
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Phosphatase 2, Regulatory Subunit 3A
- Antigen Symbol:PPP2R3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5523
- Antigen Synonyms:THIO|ACAA|PR72|PR130|PPP2R3|PTHIO
- Amino Acid Number:721 to 31
- Storage Buffer:Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
- Sequence:TCSNHEQTLSR
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 721-31 of human PPP2R3A.
- Purification:Antigen affinity purification
- Cat. No.:10733-740
- Supplier no.:PAB15655
Specifications
About this item
Goat polyclonal antibody raised aganist synthetic peptide of PPP2R3A.
Recommended Dilutions: ELISA: 1:1000
Type: Primary
Antigen: PPP2R3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0