Anti-PCGF2 Goat Polyclonal Antibody
Catalog # 10728-584
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Polycomb group ring Finger 2
- Antigen Symbol:PCGF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:7703
- Antigen Synonyms:RNF110|ZNF144|MEL-18
- Storage Buffer:Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
- Sequence:C-KMTVNGAPVPPLT
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human PCGF2.
- Purification:Antigen affinity purification
- Cat. No.:10728-584
- Supplier no.:PAB6398
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of PCGF2.
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: PCGF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human