To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Epstein-Barr virus gp 220/350
- Antigen Symbol:Epstein-Barr virus gp 220/350
- Clonality:Monoclonal
- Clone:022
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Virus
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:10 mM PBS, pH 7.2 (10 mg/mL BSA, 0.1% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified Infected B cell lysate.
- Cat. No.:10713-380
- Supplier no.:MAB5431
Specifications
About this item
Mouse monoclonal antibody raised against native Epstein-Barr virus gp 220/350.
Recommended Dilutions: Immunofluorescence: 1:15-1:50
Type: Primary
Antigen: Epstein-Barr virus gp 220/350
Clonality: FITC (Fluorescein Isothiocyanate)
Clone: Monoclonal
Conjugation: 022
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Viruses