Anti-NUPR1 Goat Polyclonal Antibody
Catalog # 10733-190
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Nuclear Protein, Transcriptional Regulator, 1
- Antigen Symbol:NUPR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:26471
- Antigen Synonyms:Nuclear protein 1|Candidate of metastasis 1|NUPR1|NUPR1_HUMAN|p8 protein|Protein p8.|COM1
- Storage Buffer:Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
- Sequence:ATFPPATSAPQQP-C
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human NUPR1.
- Purification:Antigen affinity purification
- Cat. No.:10733-190
- Supplier no.:PAB6588
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of NUPR1.
Recommended Dilutions: ELISA: 1:32000
Type: Primary
Antigen: NUPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0