Anti-Schmidtea mediterranea Dvl-b Rabbit Polyclonal Antibody
Catalog # 10732-402
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Schmidtea mediterranea Dvl-b
- Antigen Symbol:Schmidtea mediterranea Dvl-b
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus of Freshwater Flatworms Schmidtea mediterranea Dvl-b.
- Cat. No.:10732-402
- Supplier no.:PAB16048
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Schmidtea mediterranea Dvl-b.
Recommended Dilutions: ELISA: 1:2000-1:5000; Western Blot: 1 ?g/ml
Type: Primary
Antigen: Schmidtea mediterranea Dvl-b
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: