Anti-MAGEA11 Rabbit Polyclonal Antibody
Catalog # 10679-158
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Melanoma Antigen Family A11
- Antigen Symbol:MAGEA11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:4110
- Antigen Synonyms:CT1.11|MAGE-11|MAGE11|MAGEA-11
- Storage Buffer:PBS (0.09% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human MAGEA11.
- Purification:Protein G purification
- Cat. No.:10679-158
- Supplier no.:PAB4742
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MAGEA11.
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:50-100; Immunohistochemistry: 1:50-100
Type: Primary
Antigen: MAGEA11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human