Anti-TOB2 Rabbit Polyclonal Antibody
Catalog # 10680-220
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Transducer of ERBB2 2
- Antigen Symbol:TOB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:10766
- Antigen Synonyms:KIAA1663|TROB2|TOB4
- Storage Buffer:PBS (0.09% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human TOB2.
- Purification:Protein A purification
- Cat. No.:10680-220
- Supplier no.:PAB4858
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of TOB2.
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:50-100; Immunohistochemistry: 1:50
Type: Primary
Antigen: TOB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human