Anti-CXCL1 Mouse Monoclonal Antibody [clone: 5f252]
Catalog # 10709-550
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Chemokine (C-X-C Motif) Ligand 1
- Antigen Symbol:CXCL1
- Clonality:Monoclonal
- Clone:5f252
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:2919
- Antigen Synonyms:SCYB1|GROa|GRO1|MGSA-a|NAP-3|MGSA|FSP
- Amino Acid Number:36 to 107
- Storage Buffer:citrate-Tris-HCl, pH7.0 (0.02% Procl300)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 36-107 of human CXCL1.
- Purification:Affinity purification
- Cat. No.:10709-550
- Supplier no.:MAB8882
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant CXCL1.
Type: Primary
Antigen: CXCL1
Clonality: Monoclonal
Clone: 5f252
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: