To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Rabbit
- Western Blot:Yes
- Size:2 mg
- Environmentally Preferable:
- Epitope:Heavy and Light chain
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:20 mM KH2PO4, 150 mM NaCl, pH 7.2 (0.01% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Rabbit IgG whole molecule
- Purification:solid phase adsorption
- Cat. No.:10638-120
- Supplier no.:PAB9411
Sheep anti-rabbit IgG recognizes rabbit IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography.
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host:
Isotype:
Reactivity: Rabbit