Anti-CA1 Rabbit Polyclonal Antibody
10704-576
: Abnova
- Antibody Type:Primary
- Antigen Name:Carbonic Anhydrase 1
- Antigen Symbol:CA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µg
- Epitope:N-Terminal
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:759
- Antigen Synonyms:Carbonic anhydrase 1|Carbonic dehydratase|HEL-S-11|Car1|CA-I|CAB|Carbonate dehydratase I|ECK0125|yadF|Carbonic anhydrase B
- Storage Buffer:0.9 mg NaCl, 0.2 mg Na2HPO4 (5 mg BSA, 0.05 mg sodium azide, 0.05 mg Thimerosal)
- Storage Temperature:Store at –20 °C on dry atmosphere. After reconstitution with 200 ?l of deionized water and concentration will be 500 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at N-terminus of human CA1.
- Purification:Immunoaffinity purification
- Cat. No.:10704-576
- Supplier no.:PAB19518
Rabbit polyclonal antibody raised against synthetic peptide of CA1.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: CA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Rat