Anti-STARD13 Rabbit Polyclonal Antibody
Catalog # 10739-626
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:StAR-Related Lipid Transfer (START) Domain Containing 13
- Antigen Symbol:STARD13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:90627
- Antigen Synonyms:ARHGAP37|LINC00464|DLC2|GT650
- Storage Buffer:PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal of human STARD13.
- Cat. No.:10739-626
- Supplier no.:PAB17478
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of STARD13.
Recommended Dilutions: Immunofluorescence: 1:500-1:1000; ELISA: 1:10000
Type: Primary
Antigen: STARD13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse