Anti-RAF1 Rabbit Polyclonal Antibody
Catalog # 10675-250
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Raf-1 Proto-Oncogene, Serine/Threonine Kinase
- Antigen Symbol:RAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Primates,Chicken,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:24703
- Antigen Synonyms:Raf-1|CRAF|RAF|NS5|cRaf|C-raf|c-Raf|Proto-oncogene c-RAF
- Modification:phospho-Ser642
- Storage Buffer:10 mM HEPES, 150 mM NaCl, pH 7.5 (50% glycerol, 10% BSA)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S642 of rat Raf1.
- Purification:Affinity purification
- Cat. No.:10675-250
- Supplier no.:PAB9660
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Raf1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: RAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Human, Mouse, Primates, Rat