Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog # 10674-672
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Glutamate Ionotropic Receptor NMDA Type Subunit 2B
- Antigen Symbol:GRIN2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Zebrafish,Primates,Chicken,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:24410
- Antigen Synonyms:hNR3|GluN2B|NR2B|EIEE27|MRD6|NMDAR2B|glutamate ionotropic receptor NMDA type subunit 2B
- Modification:phospho-Tyr125
- Storage Buffer:10 mM HEPES, 150 mM NaCl, pH 7.5 (50% glycerol, 10% BSA)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding Y125 of rat Grin2b.
- Purification:Affinity purification
- Cat. No.:10674-672
- Supplier no.:PAB9671
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Grin2b.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: Grin2b
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Bovine, Chicken, Dog, Human, Mouse, Primates, Rat, Zebrafish