Anti-GC Goat Polyclonal Antibody
Catalog # 10715-670
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:group-specific component (vitamin D binding protein)
- Antigen Symbol:GC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Gene ID:2638
- Antigen Synonyms:VDBP|GRD3|DBP/GC|DBP|VDBG|HEL-S-51
- Storage Buffer:Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
- Sequence:CDNLSTKNSKFED
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human GC.
- Purification:Antigen affinity purification
- Cat. No.:10715-670
- Supplier no.:PAB7358
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of GC.
Recommended Dilutions: ELISA: 1:2000; Western Blot: 0.1-0.3 ?g/ml
Type: Primary
Antigen: GC
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Goat
Reactivity: Human