Anti-CACNG2 Rabbit Polyclonal Antibody
Catalog # 10705-416
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Calcium Channel, Voltage-dependent, Gamma Subunit 2
- Antigen Symbol:CACNG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Liquid
- Gene ID:84347
- Antigen Synonyms:MRD10|calcium voltage-gated channel auxiliary subunit gamma 2
- Storage Buffer:10 mM HEPES, 150 mM NaCl, pH 7.5 (50% glycerol, 10% BSA)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to C-terminus rat Cacng2.
- Purification:Affinity purification
- Cat. No.:10705-416
- Supplier no.:PAB9618
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Cacng2.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: CACNG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat