To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Glutamate Receptor 1
- Antigen Symbol:GRIA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:2890
- Antigen Synonyms:GLUH1|AMPA 1|AMPA-selective glutamate receptor 1|GLURA|GLUR1|glutamate receptor 1|gluR-A|GluA1|MGC133252|gluR-1|HBGR1|gluR-K1
- Modification:phospho-Ser849
- Storage Buffer:PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
- Sequence:Q-Q-Sp-I-N
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S849 of human GRIA1.
- Purification:purified by Affinity Chromatography
- Cat. No.:10674-660
- Supplier no.:PAB25848
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GRIA1.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: GRIA1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse, Rat