Anti-RPSAP58 Mouse Polyclonal Antibody
Catalog # 10563-208
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Ribosomal Protein SA Pseudogene 58
- Antigen Symbol:RPSAP58
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:388524
- Antigen Synonyms:RPSA_30_1642
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10563-208
- Supplier no.:H00388524-B01P
Specifications
About this item
Type: Primary
Antigen: RPSAP58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human