Anti-DNAH14 (dynein, axonemal, heavy chain 14) Mouse Monoclonal Antibody [clone: 3C12]
10593-346
: Abnova
- Antibody Type:Primary
- Antigen Name:Dynein, Axonemal, Heavy Chain 14
- Antigen Symbol:DNAH14
- Clonality:Monoclonal
- Clone:3C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:200095
- Antigen Synonyms:HL18|C1orf67|HL-18|Dnahc14
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10593-346
- Supplier no.:H00200095-M01
Type: Primary
Antigen: DNAH14 (dynein, axonemal, heavy chain 14)
Clonality: Monoclonal
Clone: 3C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human