Anti-C2ORF57 Mouse Polyclonal Antibody
10564-232
: Avantor
- Antibody Type:Primary
- Antigen Name:Chromosome 2 open reading frame 57
- Antigen Symbol:C2ORF57
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Cross Adsorption:No
- Gene ID:165100
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10564-232
- Supplier no.:H00165100-B01
Type: Primary
Antigen: C2ORF57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human